3-Amino-3-(3-methylphenyl)propionsäure
Lieferant: BIONET RESEARCH
Synonyme:
3-Amino-3-(m-tolyl)propanoic acid
7R-0654.500
7R-0654.1
7R-0654.5
BIOT7R-0654.500EA
0
EUR
BIOT7R-0654.500
BIOT7R-0654.1
BIOT7R-0654.5
3-Amino-3-(3-methylphenyl)propionsäure
3-Amino-3-(3-methylphenyl)propionsäure
Formel:
C₁₀H₁₃NO₂ Molecular Weight: 179.2157 g/mol |
MDL:
MFCD01871312 CAS-Nummer: 68208-17-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...