3-Amino-3-(4-isopropylphenyl)propionsäure
Lieferant: BIONET RESEARCH
7R-0625.500
7R-0625.1
7R-0625.5
BIOT7R-0625.500EA
0
EUR
BIOT7R-0625.500
BIOT7R-0625.1
BIOT7R-0625.5
3-Amino-3-(4-isopropylphenyl)propionsäure
3-Amino-3-(4-isopropylphenyl)propionsäure
Formel:
C₁₂H₁₇NO₂ Molecular Weight: 207,2700 g/mol |
MDL:
MFCD02663184 CAS-Nummer: 117391-53-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...