3-Amino-3-(4-ethylphenyl)propionsäure

Lieferant: BIONET RESEARCH

10R-0615.500 10R-0615.1 10R-0615.5 10R-0615.10
BIOT10R-0615.500EA 0 EUR
BIOT10R-0615.500 BIOT10R-0615.1 BIOT10R-0615.5 BIOT10R-0615.10
3-Amino-3-(4-ethylphenyl)propionsäure
3-Amino-3-(4-ethylphenyl)propionsäure
Formel: C₁₁H₁₅NO₂
MDL: MFCD01990387
CAS-Nummer: 117391-52-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR