3-Amino-3-(4-methylphenyl)propionsäure

Lieferant: BIONET RESEARCH

Synonyme: 3-Amino-3-p-tolyl-propionic acid, DL-3-Amino-3-p-tolyl-propionic acid, 3-Amino-3-(4-methylphenyl)propionic acid, 3-Amino-3-(p-tolyl)propionic acid

6R-0630.500 6R-0630.1 6R-0630.5
BIOT6R-0630.500EA 0 EUR
BIOT6R-0630.500 BIOT6R-0630.1 BIOT6R-0630.5
3-Amino-3-(4-methylphenyl)propionsäure
3-Amino-3-(4-methylphenyl)propionsäure
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,2157 g/mol
MDL: MFCD01863249
CAS-Nummer: 68208-18-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR