2-Fluor-4-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoro-4-methylbenzeneboronic acid, (2-Fluoro-4-methylphenyl)boronic acid

BB-2398-1G BB-2398-5G BB-2398-25G
COBBBB-2398-1GEA 21.6 EUR
COBBBB-2398-1G COBBBB-2398-5G COBBBB-2398-25G
2-Fluor-4-methylphenylboronsäure
2-Fluor-4-methylphenylboronsäure
Formel: C₇H₈BFO₂
Molecular Weight: 153.95 g/mol
MDL: MFCD05664239
CAS-Nummer: 170981-26-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR