2-(Difluormethoxy)benzonitril

Lieferant: Thermo Scientific

TL00767.1 TL00767.10
MAYBTL00767.1EA 0 EUR
MAYBTL00767.1 MAYBTL00767.10
2-(Difluormethoxy)benzonitril
2-(Difluormethoxy)benzonitril
Formel: C₈H₅F₂NO
Molecular Weight: 169.1282 g/mol
MDL: MFCD00221469
CAS-Nummer: 56935-78-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR