4-(Difluormethoxy)benzonitril

Lieferant: Thermo Scientific

TL00518.1 TL00518.10
MAYBTL00518.1EA 0 EUR
MAYBTL00518.1 MAYBTL00518.10
4-(Difluormethoxy)benzonitril
4-(Difluormethoxy)benzonitril
Formel: C₈H₅F₂NO
Molecular Weight: 169,1282 g/mol
MDL: MFCD00085006
CAS-Nummer: 90446-25-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR