2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolan
Lieferant: FLUOROCHEM
Synonyme:
Isopropyl pinacolyl borate, 4,4,5,5-tetramethyl-2-(propan-2-yloxy)-1,3,2-dioxaborolane
010803-100G
FL11000.POR
010803-25G
FLUO010803-100GEA
194
EUR
FLUO010803-100G
CHMPFL11000.POR
FLUO010803-25G
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolan
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolan
Formel:
C₉H₁₉BO₃ Molecular Weight: 186,06 g/mol |
MDL:
MFCD00192241 CAS-Nummer: 61676-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...