2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Isopropyl pinacolyl borate, 4,4,5,5-tetramethyl-2-(propan-2-yloxy)-1,3,2-dioxaborolane

M-Clarity™ Qualitätsstufe = MQ100

417149-100ML 417149-25ML 417149-5ML
SIAL417149-100MLEA 238 EUR
SIAL417149-100ML SIAL417149-25ML SIAL417149-5ML
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolan, Sigma-Aldrich®
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolan
Formel: C₉H₁₉BO₃
Molecular Weight: 186,06 g/mol
MDL: MFCD00192241
CAS-Nummer: 61676-62-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR