4-Methyl-1H-pyrazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Fomepizole

222569-1G 222569-5G
SIAL222569-1GEA 127 EUR
SIAL222569-1G SIAL222569-5G
4-Methyl-1H-pyrazol, Sigma-Aldrich®
4-Methyl-1H-pyrazole
Formel: C₄H₆N₂
Molecular Weight: 82,11 g/mol
MDL: MFCD00005245
CAS-Nummer: 7554-65-6
EINECS: 231-445-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR