D(-)-α-(4-Hydroxyphenyl)glycin 98+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: D(-)-4-Hydroxyphenylglycine, H-D-Phg(4-OH)-OH, (R)-(-)-2-(4-Hydroxyphenyl)glycine, (-)-4-Hydroxy-D-phenylglycine, D-(-)-p-Hydroxyphenylglycine

201320250 201321000
ACRO201320250EA 41 EUR
ACRO201320250 ACRO201321000
D(-)-α-(4-Hydroxyphenyl)glycin 98+%
D(-)-α-(4-Hydroxyphenyl)glycin
Formel: C₈H₉NO₃
Schmelzpunkt: 240 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004262
CAS-Nummer: 22818-40-2
EINECS: 245-247-7

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Specific optical rotation -155° to -161° (20°C, 589 nm) (c=1, 1N HCl)
Titration with HClO4 ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR