D(-)-α-(4-Hydroxyphenyl)glycin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
D(-)-4-Hydroxyphenylglycine, H-D-Phg(4-OH)-OH, (R)-(-)-2-(4-Hydroxyphenyl)glycine, (-)-4-Hydroxy-D-phenylglycine, D-(-)-p-Hydroxyphenylglycine
M-Clarity™ Qualitätsstufe = MQ100
215333-100G
215333-25G
SIAL215333-100GEA
38.6
EUR
SIAL215333-100G
SIAL215333-25G
D(-)-α-(4-Hydroxyphenyl)glycin, Sigma-Aldrich®
D(-)-α-(4-Hydroxyphenyl)glycin
Formel:
C₈H₉NO₃ Molecular Weight: 167.16 g/mol |
MDL:
MFCD00004262 CAS-Nummer: 22818-40-2 EINECS: 245-247-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...