5'-Brom-2'-methylacetanilid

Lieferant: COMBI-BLOCKS

Synonyme: 2-Acetamido-4-bromotoluene, N-(5-Bromo-2-methylphenyl)acetamide

HC-2832-5G HC-2832-1G
COBBHC-2832-5GEA 303 EUR
COBBHC-2832-5G COBBHC-2832-1G
5'-Brom-2'-methylacetanilid
5'-Brom-2'-methylacetanilid
Formel: C₉H₁₀BrNO
Molecular Weight: 228,0900 g/mol
MDL: MFCD11520466
CAS-Nummer: 116436-10-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR