2-Chlor-2'-methylacetanilid

Lieferant: CHEMPUR

Synonyme: 2-Chloro-N-(o-tolyl)acetamide

RH01181.250 RH01181.1
CHMPRH01181.250EA 0 EUR
CHMPRH01181.250 MAYBRH01181.1
2-Chlor-2'-methylacetanilid
2-Chlor-2'-methylacetanilid
Formel: C₉H₁₀ClNO
Molecular Weight: 183,6348 g/mol
MDL: MFCD00018894
CAS-Nummer: 37394-93-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR