2,3'-Dichlor-4'-fluoracetanilid
Lieferant: FLUOROCHEM
Synonyme:
3-Chloro-N-(chloroacetyl)-4-fluoroaniline
008223-5G
008223-25G
FLUO008223-5GEA
69.8
EUR
FLUO008223-5G
FLUO008223-25G
2,3'-Dichlor-4'-fluoracetanilid
2,3'-Dichlor-4'-fluoracetanilid
Formel:
C₈H₆Cl₂FNO |
MDL:
MFCD00084938 CAS-Nummer: 96980-64-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...