3',5'-Bis(trifluormethyl)acetanilid
Lieferant: Thermo Scientific
Synonyme:
N-(3,5-Bis(trifluoromethyl)phenyl)acetamide
TL00622.1
TL00622.10
TL00622.50
MAYBTL00622.1EA
0
EUR
MAYBTL00622.1
MAYBTL00622.10
MAYBTL00622.50
3',5'-Bis(trifluormethyl)acetanilid
3',5'-Bis(trifluormethyl)acetanilid
Formel:
C₁₀H₇F₆NO |
MDL:
MFCD00089454 CAS-Nummer: 16143-84-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...