2-Fluorisonicotinsäure

Lieferant: FLUOROCHEM

Synonyme: 2-Fluoropyridine-4-carboxylic acid

005468-500MG 005468-1G
FLUO005468-500MGEA 0 EUR
FLUO005468-500MG FLUO005468-1G
2-Fluorisonicotinsäure
2-Fluorisonicotinsäure
Formel: C₆H₄FNO₂
Molecular Weight: 141,10 g/mol
MDL: MFCD02181194
CAS-Nummer: 402-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR