6-Brom-2-naphthylamin
Lieferant: COMBI-BLOCKS
Synonyme:
6-Bromonaphthalen-2-amine, 2-Amino-6-bromonaphthalene
AN-1284-5G
AN-1284-1G
COBBAN-1284-5GEA
196
EUR
COBBAN-1284-5G
COBBAN-1284-1G
6-Brom-2-naphthylamin
6-Brom-2-naphthylamin
Formel:
C₁₀H₈BrN Molecular Weight: 222,0812 g/mol |
MDL:
MFCD01026466 CAS-Nummer: 7499-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...