Methyl-(4-bromphenyl)acetat

Lieferant: COMBI-BLOCKS

Synonyme: methyl 2-(4-bromophenyl)acetate

CA-4762-5G CA-4762-25G
COBBCA-4762-5GEA 22 EUR
COBBCA-4762-5G COBBCA-4762-25G
Methyl-(4-bromphenyl)acetat
Methyl-(4-bromphenyl)acetat
Formel: C₉H₉BrO₂
Molecular Weight: 229.0700 g/mol
MDL: MFCD00126844
CAS-Nummer: 41841-16-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR