Ethyl α-bromphenylacetat ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Ethyl alpha-bromophenylacetate
B21114.30
B21114.09
B21114.18
B21114.30EA
732
EUR
B21114.30
B21114.09
B21114.18
Ethyl α-bromphenylacetat ≥97%
Ethyl α-bromphenylacetat
Purity: 97%
CAS No.: 2882-19-1
Molecular Formula: C10H11BrO2
Formula Weight: 243.10
Flash Point:
Formel:
C₁₀H₁₁BrO₂ Siedepunkt: 164 °C (30 mmHg) Dichte: 1,389 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013536 CAS-Nummer: 2882-19-1 EINECS: 220-735-2 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...