2-Methoxy-5-methylbenzonitril, Sigma-Aldrich®

Lieferant: Merck

642908-1G 642908-5G
SIAL642908-1GEA 62.9 EUR
SIAL642908-1G SIAL642908-5G
2-Methoxy-5-methylbenzonitril, Sigma-Aldrich®
2-Methoxy-5-methylbenzonitril
Formel: C₉H₉NO
Molecular Weight: 147,1700 g/mol
MDL: MFCD03425693
CAS-Nummer: 53078-70-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR