3-Fluorsalicylaldehyd
Lieferant: FLUOROCHEM
Synonyme:
3-Fluoro-2-hydroxybenzaldehyde
006235-1G
006235-5G
FLUO006235-1GEA
36.9
EUR
FLUO006235-1G
FLUO006235-5G
3-Fluorsalicylaldehyd
3-Fluorsalicylaldehyd
Formel:
C₇H₅FO₂ Molecular Weight: 140,12 g/mol |
MDL:
MFCD00003319 CAS-Nummer: 394-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...