4-Fluorsalicylaldehyd

Lieferant: FLUOROCHEM

Synonyme: 4-Fluoro-2-hydroxybenzaldehyde

013957-1G 013957-5G
FLUO013957-1GEA 76.2 EUR
FLUO013957-1G FLUO013957-5G
4-Fluorsalicylaldehyd
4-Fluorsalicylaldehyd
Formel: C₇H₅FO₂
Molecular Weight: 140,1118 g/mol
MDL: MFCD03788526
CAS-Nummer: 348-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR