N-[2-Nitro-4-(trifluormethyl)phenyl]piperazin
Lieferant: Thermo Scientific
BTB03244DA
BTB03244EA
BTB03244.50
MAYBBTB03244DAEA
0
EUR
MAYBBTB03244DA
MAYBBTB03244EA
MAYBBTB03244.50
N-[2-Nitro-4-(trifluormethyl)phenyl]piperazin
N-[2-Nitro-4-(trifluormethyl)phenyl]piperazin
Formel:
C₁₁H₁₂F₃N₃O₂ |
MDL:
MFCD00052613 CAS-Nummer: 58315-38-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...