4,5-Dimethyl-2-isopropylthiazol

Lieferant: FLUOROCHEM

009625-1G 009625-5G
FLUO009625-1GEA 128 EUR
FLUO009625-1G FLUO009625-5G
4,5-Dimethyl-2-isopropylthiazol
4,5-Dimethyl-2-isopropylthiazol
Formel: C₈H₁₃NS
MDL: MFCD00053139
CAS-Nummer: 53498-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR