2-Chlor-6-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8579-1G BB-8579-250M
COBBBB-8579-1GEA 0 EUR
COBBBB-8579-1G COBBBB-8579-250M
2-Chlor-6-(trifluormethyl)phenylboronsäure
2-Chlor-6-(trifluormethyl)phenylboronsäure
Formel: C₇H₅BClF₃O₂
MDL: MFCD09037968
CAS-Nummer: 851756-52-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR