2,5-Dichlor-3-thiophensulfonylchlorid
Lieferant: FLUOROCHEM
CFF254.10
CFF254.25
017304-10G
017304-25G
017304-5G
CHMPCFF254.10EA
401
EUR
CHMPCFF254.10
CHMPCFF254.25
FLUO017304-10G
FLUO017304-25G
FLUO017304-5G
2,5-Dichlor-3-thiophensulfonylchlorid
2,5-Dichlor-3-thiophensulfonylchlorid
Formel:
C₄HCl₃O₂S₂ Molecular Weight: 251.54 g/mol |
MDL:
MFCD00051665 CAS-Nummer: 56946-83-9 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...