Silicium ≥99%, kristallines Pulver APS ∼100 nm, Plasmasynthese 

Lieferant: Thermo Scientific
Achtung

46308.10 46308.04 46308.09 46308.18
46308.10EA 0 EUR
46308.10 46308.04 46308.09 46308.18
Silicium ≥99%, kristallines Pulver APS ∼100 nm, Plasmasynthese 
Silicium

Purity: 99% CAS No.: 7440-21-3 Odour: Odourless UN No.: 1346 MDL No.: MFCD00085311

Formel: Si
Molecular Weight: 28,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00085311
CAS-Nummer: 7440-21-3
EINECS: 231-130-8
UN: 1346
ADR: 4.1,III
Merck Index: 12,08635

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR