Ethylphenylcyanacetat ≥98%
Lieferant: Thermo Scientific
L03929.06
L03929.14
L03929.06EA
32.7
EUR
L03929.06
L03929.14
Ethylphenylcyanacetat ≥98%
Ethylphenylcyanacetat
Purity: 98%
CAS No.: 4553-07-5
Molecular Formula: C11H11NO2
Formula Weight: 189.21
Flash Point:
Formel:
C₁₁H₁₁NO₂ Molecular Weight: 189,21 g/mol Siedepunkt: 274…275 °C Dichte: 1,090 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001866 CAS-Nummer: 4553-07-5 EINECS: 224-921-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...