4-Brom-2,6-dichloranisol
Lieferant: COMBI-BLOCKS
Synonyme:
5-Bromo-1,3-dichloro-2-methoxybenzene
OT-0813-5G
OT-0813-1G
COBBOT-0813-5GEA
595
EUR
COBBOT-0813-5G
COBBOT-0813-1G
4-Brom-2,6-dichloranisol
4-Brom-2,6-dichloranisol
Formel:
C₇H₅BrCl₂O Molecular Weight: 255,9240 g/mol |
MDL:
MFCD10699182 CAS-Nummer: 19240-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...