4-Brom-2-fluorthioanisol
Lieferant: COMBI-BLOCKS
Synonyme:
(4-Bromo-2-fluorophenyl)(methyl)sulphane
OT-0473-5G
OT-0473-1G
COBBOT-0473-5GEA
449
EUR
COBBOT-0473-5G
COBBOT-0473-1G
4-Brom-2-fluorthioanisol
4-Brom-2-fluorthioanisol
Formel:
C₇H₆BrFS Molecular Weight: 221,0899 g/mol |
MDL:
MFCD12756455 CAS-Nummer: 221030-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...