5-Formylsalicylsäure
Lieferant: FLUOROCHEM
Synonyme:
5-Formyl-2-hydroxybenzoic acid
013893-5G
013893-1G
FLUO013893-5GEA
48.8
EUR
FLUO013893-5G
FLUO013893-1G
5-Formylsalicylsäure
5-Formylsalicylsäure
Formel:
C₈H₆O₄ Molecular Weight: 166,1308 g/mol |
MDL:
MFCD00006945 CAS-Nummer: 616-76-2 EINECS: 210-492-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...