Isobutyl-4-hydroxybenzoat ≥98%
Lieferant: Thermo Scientific
Synonyme:
Isobutyl 4-hydroxybenzoate, Isobutylparaben
L17587.06
L17587.18
L17587.06EA
30.2
EUR
L17587.06
L17587.18
Isobutyl-4-hydroxybenzoat ≥98%
Isobutyl-4-hydroxybenzoat
Purity: 98%
CAS No.: 4247-02-3
Molecular Formula: C11H14O3
Formula Weight: 194.23
Melting Point: 74-78°
Beilstein Reference: 2642305
Formel:
C₁₁H₁₄O₃ Molecular Weight: 194,23 g/mol Schmelzpunkt: 74…78 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00020167 CAS-Nummer: 4247-02-3 EINECS: 224-208-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...