4-Methyl-2-nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4,4,5,5-Tetramethyl-2-(4-methyl-2-nitrophenyl)-1,3,2-dioxaborolane

PN-2147-1G PN-2147-250M
COBBPN-2147-1GEA 742 EUR
COBBPN-2147-1G COBBPN-2147-250M
4-Methyl-2-nitrophenylboronsäurepinakolester
4-Methyl-2-nitrophenylboronsäurepinakolester
Formel: C₁₃H₁₈BNO₄
Molecular Weight: 263,100 g/mol
MDL: MFCD06795682
CAS-Nummer: 1256359-10-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR