3,5-Dichlor-4-isopropoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3,5-Dichloro-4-isopropoxyphenyboronic acid

BB-6231-5G BB-6231-1G
COBBBB-6231-5GEA 884 EUR
COBBBB-6231-5G COBBBB-6231-1G
3,5-Dichlor-4-isopropoxyphenylboronsäure
3,5-Dichlor-4-isopropoxyphenylboronsäure
Formel: C₉H₁₁BCl₂O₃
MDL: MFCD15143466
CAS-Nummer: 1218790-62-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR