3,5-Dichlor-4-ethoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5538-5G BB-5538-1G
COBBBB-5538-5GEA 0 EUR
COBBBB-5538-5G COBBBB-5538-1G
3,5-Dichlor-4-ethoxyphenylboronsäure
3,5-Dichlor-4-ethoxyphenylboronsäure
Formel: C₈H₉BCl₂O₃
MDL: MFCD08701776
CAS-Nummer: 1107604-10-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR