3,5-Dichlor-4-propoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-6230-5G BB-6230-1G
COBBBB-6230-5GEA 451 EUR
COBBBB-6230-5G COBBBB-6230-1G
3,5-Dichlor-4-propoxyphenylboronsäure
3,5-Dichlor-4-propoxyphenylboronsäure
Formel: C₉H₁₁BCl₂O₃
MDL: MFCD15143465
CAS-Nummer: 1218790-61-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR