3,5-Difluor-4-hydroxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3,5-Difluoro-2-hydroxyphenyl)boronic acid

BB-5623-1G BB-5623-250M
COBBBB-5623-1GEA 764 EUR
COBBBB-5623-1G COBBBB-5623-250M
3,5-Difluor-4-hydroxyphenylboronsäure
3,5-Difluor-4-hydroxyphenylboronsäure
Formel: C₆H₅BF₂O₃
Molecular Weight: 173.91 g/mol
MDL: MFCD03095356
CAS-Nummer: 1150114-51-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR