3-Isopropylphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(3-Isopropylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Isopropylphenylboronic acid, pinacol ester

PN-2133-5G PN-2133-1G
COBBPN-2133-5GEA 1320 EUR
COBBPN-2133-5G COBBPN-2133-1G
3-Isopropylphenylboronsäurepinakolester
3-Isopropylphenylboronsäurepinakolester
Formel: C₁₅H₂₃BO₂
Molecular Weight: 246,1529 g/mol
MDL: MFCD13195750
CAS-Nummer: 325142-89-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR