4-Isopropylphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-(4-Isopropylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PN-2132-1G
PN-2132-5G
COBBPN-2132-1GEA
181
EUR
COBBPN-2132-1G
COBBPN-2132-5G
4-Isopropylphenylboronsäurepinakolester
4-Isopropylphenylboronsäurepinakolester
Formel:
C₁₅H₂₃BO₂ Molecular Weight: 246,1529 g/mol |
MDL:
MFCD15143592 CAS-Nummer: 325142-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...