4-Brombenzylmethylether, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Bromo-4-(methoxymethyl)benzene

680419-10G 680419-1G
SIAL680419-10GEA 172 EUR
SIAL680419-10G SIAL680419-1G
4-Brombenzylmethylether, Sigma-Aldrich®
4-Brombenzylmethylether
Formel: C₈H₉BrO
Molecular Weight: 201,0600 g/mol
MDL: MFCD09265134
CAS-Nummer: 1515-88-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR