Benzylchloromethylether, Sigma-Aldrich®

Lieferant: Merck

Synonyme: ((Chloromethoxy)methyl)benzene

13282-10ML 13282-50ML
SIAL13282-10MLEA 50.7 EUR
SIAL13282-10ML SIAL13282-50ML
Benzylchloromethylether, Sigma-Aldrich®
Benzylchloromethylether
Formel: C₈H₉ClO
Molecular Weight: 156,61 g/mol
MDL: MFCD00000886
CAS-Nummer: 3587-60-8
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR