4,5-Difluor-2-nitrotoluol

Lieferant: CHEMPUR

Synonyme: 1,2-Difluoro-4-methyl-5-nitrobenzene

FL13822.POR 013822-5G 013822-1G
CHMPFL13822.POREA 0 EUR
CHMPFL13822.POR FLUO013822-5G FLUO013822-1G
4,5-Difluor-2-nitrotoluol
4,5-Difluor-2-nitrotoluol
Formel: C₇H₅F₂NO₂
Molecular Weight: 173,12 g/mol
MDL: MFCD04973772
CAS-Nummer: 127371-50-0
UN: 2810
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR