2,4-Dinitro-5-fluortoluol

Lieferant: FLUOROCHEM

Synonyme: 1-Fluoro-5-methyl-2,4-dinitrobenzene

004969-1G 004969-5G
FLUO004969-1GEA 85.1 EUR
FLUO004969-1G FLUO004969-5G
2,4-Dinitro-5-fluortoluol
2,4-Dinitro-5-fluortoluol
Formel: C₇H₅FN₂O₄
Molecular Weight: 200.1240 g/mol
MDL: MFCD00205111
CAS-Nummer: 349-01-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR