1-Brom-3,5-di-tert-butylbenzol ≥99%

Lieferant: Thermo Fisher Scientific

L14646.03 L14646.06 L14646.14
L14646.03EA 52.6 EUR
L14646.03 L14646.06 L14646.14
1-Brom-3,5-di-tert-butylbenzol ≥99%
1-Brom-3,5-di-tert-butylbenzol

Purity: 99% CAS No.: 22385-77-9 Molecular Formula: C14H21Br Formula Weight: 269.23 Melting Point: 62-66° Beilstein Reference: 2091559

Formel: C₁₄H₂₁Br
Molecular Weight: 269.23 g/mol
Schmelzpunkt: 62…66 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00796945
CAS-Nummer: 22385-77-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR