1-Brom-3,5-di-tert-butylbenzol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

592161-10G 592161-1G
SIAL592161-10GEA 251 EUR
SIAL592161-10G SIAL592161-1G
1-Brom-3,5-di-tert-butylbenzol, Sigma-Aldrich®
1-Brom-3,5-di-tert-butylbenzol
Formel: C₁₄H₂₁Br
Molecular Weight: 269.23 g/mol
MDL: MFCD00796945
CAS-Nummer: 22385-77-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR