1-Brom-4-(trans-4-pentylcyclohexyl)benzol

Lieferant: COMBI-BLOCKS

Synonyme: 1-Bromo-4-(trans-4-N-pentylcyclohexyl)benzene

OS-7498-5G OS-7498-25G OS-7498-1G
COBBOS-7498-5GEA 35.9 EUR
COBBOS-7498-5G COBBOS-7498-25G COBBOS-7498-1G
1-Brom-4-(trans-4-pentylcyclohexyl)benzol
1-Bromo-4-(trans-4-pentylcyclohexyl)benzene
Formel: C₁₇H₂₅Br
Molecular Weight: 309,2800 g/mol
MDL: MFCD00577359
CAS-Nummer: 79832-89-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR