1-Brom-4-(trans-4-propylcyclohexyl)benzol

Lieferant: COMBI-BLOCKS

OS-7497-1G OS-7497-25G OS-7497-5G
COBBOS-7497-1GEA 26.2 EUR
COBBOS-7497-1G COBBOS-7497-25G COBBOS-7497-5G
1-Brom-4-(trans-4-propylcyclohexyl)benzol
1-Bromo-4-(trans-4-propylcyclohexyl)benzene
Formel: C₁₅H₂₁Br
Molecular Weight: 281,2300 g/mol
MDL: MFCD00577367
CAS-Nummer: 86579-53-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR