Methyl-3,5-dibromsalicylat ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl 3,5-dibromo-2-hydroxybenzoate

B25077.14 B25077.22
B25077.14EA 44.9 EUR
B25077.14 B25077.22
Methyl-3,5-dibromsalicylat ≥98%
Methyl-3,5-dibromsalicylat

Purity: 98% CAS No.: 21702-79-4 Molecular Formula: C8H6Br2O3 Formula Weight: 309.94 Melting Point: 150-152° MDL No.: MFCD00017534

Formel: C₈H₆Br₂O₃
Molecular Weight: 309,94 g/mol
Schmelzpunkt: 150…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017534
CAS-Nummer: 21702-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR