Methyl-4-brom-3-hydroxybenzoat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 4-bromo-3-hydroxybenzoate
CA-4189-5G
CA-4189-1G
COBBCA-4189-5GEA
210
EUR
COBBCA-4189-5G
COBBCA-4189-1G
Methyl-4-brom-3-hydroxybenzoat
Methyl-4-brom-3-hydroxybenzoat
Formel:
C₈H₇BrO₃ Molecular Weight: 231,0400 g/mol |
MDL:
MFCD08056372 CAS-Nummer: 106291-80-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...